G2767
Glass microcarrier beads
150-210 μm, density: 1.03 g/cm3 at 25 °C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
41100000
Recommended Products
material
glass
particle size
150-210 μm
density
1.03 g/cm3 at 25 °C
Looking for similar products? Visit Product Comparison Guide
Application
For suspension culture of anchorage-dependent cells.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service