G2388
Globulins Cohn fraction II, III human
lyophilized powder
Synonym(s):
Human globulins, Powder globulins
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
form
lyophilized powder
composition
Protein, 60-85% TCA-Biuret
concentration
60-85% (biuret)
impurities
HIV 1 and 2, HBsAg and HCV, source material tested negative
storage temp.
2-8°C
Application
Globulins Cohn fraction II, III human has been used to replace α and β subunits to determine the role of α and β subunits of phosphorylase kinase.
Other Notes
Primarily γ- and β-globulins
Physical form
Contains NaCl
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Skin pharmacology : the official journal of the Skin Pharmacology Society, 6(1), 45-51 (1993-01-01)
The binding of 5-methoxypsoralen (5-MOP) to human serum and blood fractions was studied by equilibrium dialysis associated to high-performance liquid chromatography. 5-MOP serum binding was 95% and kept constant in the range of therapeutic concentrations. Albumin was the main binding
Inhibition of the catalytic subunit of phosphorylase kinase by its alpha/beta subunits.
The Journal of biological chemistry, 262(25), 11912-11915 (1987)
International journal of laboratory hematology, 32(1 Pt 1), e96-105 (2009-07-15)
Immune humoral neutropenia (Np) could be the consequence of anti-polymorphonuclear neutrophil (PMN) antibodies, circulating immune complexes (CIC) and/or antibodies against myeloid precursors. Granulocyte immunofluorescence test (GIFT) and a leukoagglutination technique (LAGT) assays are recommended for its diagnosis. Fifty adult patients
ALTEX, 37(3), 451-468 (2020-04-24)
There are currently no skin sensitization assays based on T cell activation. We built a novel in vitro model to assess T cell activation and test its performance to discriminate skin sensitizers from non-sensitizers using 52 reference chemicals. Jurkat Clone
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service