Skip to Content
Merck
All Photos(4)

Key Documents

G2140

Sigma-Aldrich

L-Glutathione oxidized disodium salt

BioReagent, suitable for cell culture

Synonym(s):

GSSG

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$115.00
1 G
$348.00
5 G
$1,230.00

$115.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$115.00
1 G
$348.00
5 G
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C20H30N6Na2O12S2
CAS Number:
Molecular Weight:
656.59
Beilstein:
6265389
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75

$115.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

biological source

yeast

Quality Level

product line

BioReagent

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

ethanol, essentially free

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

[Na+].[Na+].N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C([O-])=O)C(=O)NCC(O)=O)C(=O)NCC([O-])=O)C(O)=O

InChI

1S/C20H32N6O12S2.2Na/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38;;/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38);;/q;2*+1/p-2/t9-,10-,11-,12-;;/m0../s1

InChI key

DRFILBXQKYDTFW-JIWRMXRASA-L

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

(γ-Glu-Cys-Gly) (γ-Glu-Cys-Gly) [Disulfide bridge: 2a-2b]

Application

For use in cell culture applications as an antioxidant and heavy metal detoxification agent. L-Glutathione oxidized disodium salt has been used in semen processing and evaluation.[1]

Biochem/physiol Actions

Oxidized L-glutathione is frequently measured in vivo as an indicator of oxidative stress.[2]

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Influence of various antioxidants on microscopic-oxidative stress indicators and fertilizing ability of frozen-thawed bull semen
Sariozkan S, et al.
Acta Veterinaria Brno, 78(3), 463-469 (2009)
Oxidized forms of glutathione in peripheral blood as biomarkers of oxidative stress.
Rossi R
Clinical Chemistry, 52(7), 1406-1414 (2006)
Christian Stoppe et al.
Antioxidants & redox signaling, 23(11), 865-879 (2015-08-04)
Cardiac surgery involves myocardial ischemia/reperfusion (I/R) with potentially deleterious consequences. Macrophage migration inhibitory factor (MIF) is a stress-regulating chemokine-like cytokine that protects against I/R damage, but functional links with its homolog, d-dopachrome tautomerase (MIF-2), and the circulating soluble receptor CD74
Katia M S Gomes et al.
International journal of cardiology, 179, 129-138 (2014-12-03)
We previously demonstrated that reducing cardiac aldehydic load by aldehyde dehydrogenase 2 (ALDH2), a mitochondrial enzyme responsible for metabolizing the major lipid peroxidation product, protects against acute ischemia/reperfusion injury and chronic heart failure. However, time-dependent changes in ALDH2 profile, aldehydic
Haitao Shi et al.
Journal of experimental botany, 66(3), 681-694 (2014-09-17)
Melatonin (N-acetyl-5-methoxytryptamine), a well-known animal hormone, is also involved in plant development and abiotic stress responses. In this study, it is shown that exogenous application of melatonin conferred improved salt, drought, and cold stress resistances in bermudagrass. Moreover, exogenous melatonin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service