Skip to Content
Merck
All Photos(1)

Documents

G212050

Sigma-Aldrich

DMT-2′O-TBDMS-rG(tac) Phosphoramidite

configured for ABI

Synonym(s):

DMT-2′-O-TBDMS-rG(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine,3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H76N7O10PSi
CAS Number:
Molecular Weight:
1090.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

compatibility

configured for ABI

nucleoside profile

base: guanosine
base protecting group: TAC
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)COc5ccc(cc5)C(C)(C)C)=Nc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C58H76N7O10PSi/c1-38(2)65(39(3)4)76(72-34-18-33-59)74-50-47(35-71-58(41-19-16-15-17-20-41,42-23-27-44(68-11)28-24-42)43-25-29-45(69-12)30-26-43)73-54(51(50)75-77(13,14)57(8,9)10)64-37-60-49-52(64)62-55(63-53(49)67)61-48(66)36-70-46-31-21-40(22-32-46)56(5,6)7/h15-17,19-32,37-39,47,50-51,54H,18,34-36H2,1-14H3,(H2,61,62,63,66,67)/t47-,50-,51-,54-,76?/m1/s1

InChI key

ODCBQUOWLVQKQG-PZUNXSRFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service