Skip to Content
Merck
All Photos(1)

Key Documents

G1642

Sigma-Aldrich

sn-Glycerol-3-phosphocholine Phosphodiesterase from mold

lyophilized powder, ≥5 units/mg protein

Synonym(s):

Glycerophosphorylcholine phosphodiesterase from mold

Sign Into View Organizational & Contract Pricing

Select a Size

10 UNITS
$590.00

$590.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
10 UNITS
$590.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$590.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


form

lyophilized powder

Quality Level

specific activity

≥5 units/mg protein

composition

Protein, ~40% Bradford

storage temp.

−20°C

Unit Definition

One unit will produce 1.0 μmole of choline from L-α-glycerophosphorylcholine, G4007, per min at pH 8.0 at 37 °C.

Physical form

Lyophilized powder containing Tris buffer salt

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sipione et al.
FEBS letters, 384(1), 19-24 (1996-04-08)
In microvessels isolated from bovine brain, microsomal enzyme activities involved in phosphatidylcholine biosynthesis and degradation were determined. The microvessels possessed acyl-CoA:1-acyl-sn-glycero-3-phosphocholine (AT) and glycerophosphocholine phosphodiesterase (GroPChoPDE) activity at a higher level compared with bovine and rat brain or rat liver
Carmelina D Anfuso et al.
Lipids, 38(1), 45-52 (2003-04-03)
In pericytes from bovine retina, the enzyme glycerophosphocholine phosphodiesterase, catalyzing the hydrolysis of sn-glycero-3-phosphocholine to glycero-3-phosphate and choline, has been characterized with respect to pH optimum, metal ion dependence, Km, inhibitors, and subcellular localization. In these cells, the natural substrate
Mária Simocková et al.
The Journal of biological chemistry, 283(25), 17107-17115 (2008-04-25)
The product of the open reading frame YPL206c, Pgc1p, of the yeast Saccharomyces cerevisiae displays homology to bacterial and mammalian glycerophosphodiester phosphodiesterases. Deletion of PGC1 causes an accumulation of the anionic phospholipid, phosphatidylglycerol (PG), especially under conditions of inositol limitation.
J N Kanfer et al.
Neurochemical research, 18(3), 331-334 (1993-03-01)
Previous studies have demonstrated elevated brain levels of phosphomonoesters in early stages of Alzheimer's disease and elevations of phosphodiesters later in the disease. In addition, preliminary quantitative analyses of the phospholipids of Alzheimer's brain reveals either decreases in some phospholipids
J P Galons et al.
Magnetic resonance in medicine, 33(3), 422-426 (1995-03-01)
The purpose of this study was to study the metabolic events during a slow acidosis in three different cell lines by combining 31P magnetic resonance spectroscopy and hollow fiber bioreactor technology. The rate of change in intracellular pH, glycerophosphorylcholine (GPC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service