Skip to Content
Merck
All Photos(1)

Documents

G1506

Sigma-Aldrich

Guanylyl(3′→5′)cytidine ammonium salt

Synonym(s):

GpC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H25N8O12P · NH3
CAS Number:
Molecular Weight:
605.45
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

N.NC1=NC(=O)c2ncn(C3OC(CO)C(OP(O)(=O)OCC4OC(C(O)C4O)N5C=CC(N)=NC5=O)C3O)c2N1

InChI

1S/C19H25N8O12P.H3N/c20-8-1-2-26(19(33)23-8)16-11(30)10(29)7(38-16)4-36-40(34,35)39-13-6(3-28)37-17(12(13)31)27-5-22-9-14(27)24-18(21)25-15(9)32;/h1-2,5-7,10-13,16-17,28-31H,3-4H2,(H,34,35)(H2,20,23,33)(H3,21,24,25,32);1H3

InChI key

PPSONIOCBRREER-UHFFFAOYSA-N

Application

Guanylyl(3′5′)cytidine is an oligonucleotide pyrimidine dimer used as a substrate to study the specificity and kinetics of various RNases such as RNase T1 and its mutations.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Vincent et al.
Genome biology, 18(1), 38-38 (2017-02-23)
The study of epigenetic heterogeneity at the level of individual cells and in whole populations is the key to understanding cellular differentiation, organismal development, and the evolution of cancer. We develop a statistical method, epiG, to infer and differentiate between
Rico Czaja et al.
Biochemistry, 43(10), 2854-2862 (2004-03-10)
Attempts to alter the guanine specificity of ribonuclease T1 (RNase T1) by rational or random mutagenesis have failed so far. The RNase T1 variant RV (Lys41Glu, Tyr42Phe, Asn43Arg, Tyr45Trp, and Glu46Asn) designed by combination of a random and a rational
Marc Struhalla et al.
Chembiochem : a European journal of chemical biology, 5(2), 200-205 (2004-02-05)
Although ribonuclease T1 (RNase T1) is one of the best-characterized proteins with respect to structure and enzymatic action, numerous attempts at altering the specificity of the enzyme to cleave single-stranded RNA at the 3'-side of adenylic instead of guanylic residues
Alexandra Søgaard Helbo et al.
Scientific reports, 7, 41947-41947 (2017-02-09)
Chromatin is structurally involved in the transcriptional regulation of all genes. While the nucleosome positioning at RNA polymerase II (pol II) promoters has been extensively studied, less is known about the chromatin structure at pol III promoters in human cells.
Snawar Hussain et al.
Acta tropica, 173, 136-146 (2017-06-14)
Zika virus is a zoonotic pathogen, which have made frequent incursion into the human population in Africa and South East Asia over the course of several decades but never reached to the pandemic proportions until the most recent outbreak. Viruses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service