Skip to Content
Merck
All Photos(1)

Key Documents

G135

Sigma-Aldrich

Guanosine 3′,5′-cyclic monophosphorothioate, Rp Isomer triethylammonium salt

solid

Synonym(s):

Rp-Cyclic 3′,5′-hydrogen phosphorothioate guanosine triethylammonium salt, Rp-Diastereomer of guanosine 3′,5′-cyclic monophosphorothioate triethylammonium salt, Rp-cGMPS triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5O6PS · C6H15N
CAS Number:
Molecular Weight:
462.46
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: 5 mg/mL

storage temp.

−20°C

SMILES string

CCN(CC)CC.NC1=Nc2c(ncn2[C@@H]3O[C@@H]4CO[P@@](O)(=S)O[C@H]4[C@H]3O)C(=O)N1

InChI

1S/C10H12N5O6PS.C6H15N/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(20-9)1-19-22(18,23)21-6;1-4-7(5-2)6-3/h2-3,5-6,9,16H,1H2,(H,18,23)(H3,11,13,14,17);4-6H2,1-3H3/t3-,5-,6-,9-,22-;/m1./s1

InChI key

SBVIQYPGQBMDDI-FBGJTXKBSA-N

Biochem/physiol Actions

cGMPS-dependent protein kinase Iα inhibitor.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nardhy Gomez-Lopez et al.
Cell reports, 32(1), 107874-107874 (2020-07-09)
Regulatory T cells (Tregs) have been exhaustively investigated during early pregnancy; however, their role later in gestation is poorly understood. Herein, we report that functional Tregs are reduced at the maternal-fetal interface in a subset of women with idiopathic preterm labor/birth
E Butt et al.
FEBS letters, 263(1), 47-50 (1990-04-09)
The activation of the cGMP-dependent protein kinase and cAMP-dependent protein kinase by the diastereomers of guanosine 3',5'-monophosphorothioate, (Sp)-cGMPS and (Rp)-cGMPS, and 8-chloroguanosine 3',5'-monophosphorothioate, (Sp)-8-Cl-cGMPS and (Rp)-8-Cl-cGMPS, was investigated using the peptide Kemptide as substrate. The (Sp)-diastereomers, which have an axial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service