Skip to Content
Merck
All Photos(1)

Key Documents

G1270

Sigma-Aldrich

L-Glutamine Synthetase from Escherichia coli

lyophilized powder, 400-2,000 units/mg protein

Synonym(s):

L-Glutamate:ammonia ligase (ADP-forming)

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
$851.00

$851.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
100 UNITS
$851.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

$851.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

form

lyophilized powder

Quality Level

specific activity

400-2,000 units/mg protein

purified by

affinity chromatography

contains

dithioerythritol as preservative

composition

Protein, ~5% Lowry

solubility

H2O: soluble 0.95-1.05 mg/mL, clear to hazy

UniProt accession no.

foreign activity

ATPase <0.2%

storage temp.

−20°C

Gene Information

Escherichia coli K12 ... glnA(948370)

General description

L-Glutamine Synthetase from bacteria shows dodecameric structure comprising of 12 active sites. Each active site termed bifunnel, has an ATP and glutamate binding sites. The dodecamer is stabilized by two hexameric rings.[1]

Application

L-Glutamine Synthetase from Escherichia coli has been used in the synthesis of methylglutamine from methylammonium in E coli[2] and in the glutamine synthetase protection activity of human thioredoxin peroxidase enzyme, AOE372.[3]
L-Glutamine synthetase may be used for the purification of proteases from Escherichia coli.[4]

Biochem/physiol Actions

L-glutamine synthetase catalyzes the condensation of L-glutamate and ammonia to L-glutamine[5]. It is a degradative enzyme for glutamic acid.
Nitrogen starvation dictates the expression of the glutamine synthetase (GS) gene in E. coli.[1] GS plays a key role in ammonia assimilation in bacteria.[6] Adenylylation of GS is catalyzed by adenylyltransferase. Adenylylation of GS modulates its catalytic functionality resulting in glutamine limitation in E coli.[7]
Degradative enzyme for glutamic acid

Unit Definition

One unit will convert 1.0 μmole of L-glutamate to L-glutamine in 15 min at pH 7.1 at 37 °C.

Physical form

Contains potassium phosphate, sodium citrate and magnesium acetate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulatory role for a novel human thioredoxin peroxidase in NF-kappaB activation
Jin DY, et al.
The Journal of Biological Chemistry, 272(49), 30952-30961 (1997)
Reversible Adenylylation of Glutamine Synthetase Is Dynamically Counterbalanced during Steady-State
Okano H, et al.
Journal of molecular biology, 404(1), 522-536 (2010)
Huijuan Jia et al.
Molecular nutrition & food research, 57(2), 291-306 (2012-11-21)
This study addresses the effects of branched-chain amino acids (BCAA) on global gene expression in liver and skeletal muscle and the molecular mechanisms underlying the improvement in liver cirrhosis using DNA microarray analysis combined with RNase protection assay. Male Wistar
Jane E Ladner et al.
Biochemistry, 51(51), 10121-10123 (2012-12-14)
The structure of PA5508 from Pseudomonas aeruginosa, a glutamine synthetase (GS) homologue, has been determined at 2.5 Å. Surprisingly, PA5508 forms single hexameric rings rather than the stacked double rings that are characteristic of GS. The C-terminal helical thong motif
Peng Jiang et al.
Biochemistry, 51(45), 9032-9044 (2012-10-24)
Uridylyltransferase/uridylyl-removing enzyme (UTase/UR) catalyzes uridylylation of PII and deuridylylation of PII-UMP, with both activities regulated by glutamine. In a reconstituted UTase/UR-PII cycle containing wild-type UTase/UR, the steady-state modification of PII varied from nearly complete modification to nearly complete demodification as

Questions

  1. From which E. coli strain is your product G1270 L-Glutamine Synthetase from Escherichia coli isolated?

    1 answer
    1. The K12 strain information can be found on the UniProt website under the following entry: P0A9C5.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service