Skip to Content
Merck
All Photos(1)

Key Documents

G10

Supelco

Spectrophotometer cuvettes, glass

Without stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

glass

parameter

320-1,000 nm spectral range

H × W × D

45 mm × 12.5 mm × 12.5 mm

chamber volume

≥3 mL

pathlength

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

See transmission curve labelled E

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rondine J Allen et al.
Molecular pharmaceutics, 15(9), 3881-3891 (2018-07-28)
PEGylated polylysine peptides represent a new class of scavenger receptor inhibitors that may find utility at inhibiting DNA nanoparticle uptake by Kupffer cells in the liver. PEG-peptides inhibit scavenger receptors in the liver by a novel mechanism involving in situ
Dongxi Xiang et al.
Theranostics, 7(17), 4071-4086 (2017-11-22)
Chemotherapy-resistant cancer stem cells (CSCs) are a major obstacle to the effective treatment of many forms of cancer. To overcome CSC chemo-resistance, we developed a novel system by conjugating a CSC-targeting EpCAM aptamer with doxorubicin (Apt-DOX) to eliminate CSCs. Incubation
Masanori Sakamoto et al.
Nature communications, 10(1), 406-406 (2019-01-27)
Infrared-light-induced carrier transfer is a key technology for 'invisible' optical devices for information communication systems and energy devices. However, clear and colourless photo-induced carrier transfer has not yet been demonstrated in the field of photochemistry, to the best of our
P Abate et al.
Neuropeptides, 62, 45-56 (2016-11-28)
Several studies suggest that prenatal ethanol exposure (PEE) facilitates ethanol intake. Opioid peptides play a main role in ethanol reinforcement during infancy and adulthood. However, PEE effects upon motor responsiveness elicited by an ethanol challenge and the participation of opioids
Elizabeth M Rhea et al.
Journal of Alzheimer's disease : JAD, 57(1), 241-252 (2017-02-23)
Insulin delivered to the level of the cribriform plate (intranasal insulin) is being investigated for its ability to enhance memory in people with Alzheimer's disease (AD). Recent work has shown intranasal insulin can be detected in young CD-1 mice within

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service