Skip to Content
Merck
All Photos(1)

Key Documents

G0897

Sigma-Aldrich

Genistin

from Glycine max (soybean), ≥95% (HPLC)

Synonym(s):

4′,5,7-Trihydroxyisoflavone 7-glucoside, Genistein 7-glucoside, Genistein-7-O-β-D-glucopyranoside, Genistoside, NSC 5112

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$101.00
5 MG
$353.00

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$101.00
5 MG
$353.00

About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
64479
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

Glycine max (soybean)

Quality Level

Assay

≥95% (HPLC)

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C(=COc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-5-13(24)16-14(6-11)29-8-12(17(16)25)9-1-3-10(23)4-2-9/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1

InChI key

ZCOLJUOHXJRHDI-CMWLGVBASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Genistin has been used for immune reactivity against influenza viruses in vitro[1] and as an internal standard in isoflavones quantification.[2]

Biochem/physiol Actions

Inactive analog of genistein; useful as a negative control for genistein and other tyrosine kinase inhibitors.
Selective inhibitor of mammalian terminal deoxynucleotidyl transferase (TdT), with no measurable effect on mammalian or microbial DNA polymerases.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism and Histamine Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Hamakawa et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(21), 9306-9312 (1999-10-26)
Neurotrophic factors have well established roles in neuronal development and adult synaptic plasticity, but their precise role in synapse formation has yet to be determined. This paper provides the first direct evidence that neurotrophic factors in brain conditioned medium (CM)
Soy saponins meditate the progression of colon cancer in rats by inhibiting the activity of beta-glucuronidase and the number of aberrant crypt foci but not cyclooxygenase-2 activity.
Guo YW, et al.
ISRN Oncology, 2013 (2013)
Anna Hsu et al.
Experimental biology and medicine (Maywood, N.J.), 235(1), 90-97 (2010-04-21)
Previous studies have suggested that soy isoflavones exert anticarcinogenic effects against prostate cancer. We propose that soy extracts, containing a mixture of soy isoflavones and other bioactive components, would be a more potent chemo-preventive agent than individual soy isoflavones. We
Soha M Hamdy et al.
Toxicology and industrial health, 28(8), 746-757 (2011-11-18)
Breast cancer is the second leading cause of cancer death among women and the third most common cancer. In this study, we investigated the chemoprevention efficacy of each of soy genistin, selenium or a combination of them against breast cancer.
Zong-Ping Zheng et al.
Food & function, 2(5), 259-264 (2011-07-23)
The phytochemcal profiles of Cudrania cochinchinensis leaf, twig, stem and root were compared by HPLC analysis. It was found that C. cochinchinensis stem extract contained some unknown natural products with potential tyrosinase inhibitory activities. Therefore, the chemical constitutes in extract

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Questions

1–2 of 2 Questions  
  1. What is the lambda max for genistin and what is the molar extinction coefficient at that wavelength?

    1 answer
    1. We have data from our QC labs here at Sigma on one of the Genistin products we offer, G0897. The solvent is 85% ethanol/15% water, and the lambda max is typically 262.0 or 262.5 nm. We measured the molar extinction coefficients at the lambda max for four different lots of G0897 back in the 1990s, and the average value was 39,800 M-1.Genistin is a derivative of Genistein. There is an entry for Genistein in the chemicals encyclopedia published by the Royal Society of Chemistry, with information for Genistin under that heading.According to the chemicals encyclopedia published by the Royal Society of Chemistry, 14th edition, entry #4391, uv max (85% ethanol) 262.5 nm (a 90.5)".The value of ""a 90.5"" is the same as an E0.1%, or the extinction coefficient for a 1 mg/mL (or 1 gram per Liter) solution.If one multiplies 90.5 (L/g) by the molecular weight of genistin (432.38 g/mole), you get 39,130 M-1. This is pretty close to the value of 39,800 from our data above."

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service