Skip to Content
Merck
All Photos(1)

Key Documents

FG1215

Sigma-Aldrich

RPMI-1640 Medium

With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$42.80

$42.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$42.80

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$42.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Product Name

RPMI-1640 Medium, With L-alanyl-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

sodium pyruvate: no
NaHCO3: yes
HEPES: no
phenol red: yes
stable glutamine: yes

shipped in

ambient

storage temp.

2-8°C

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells.

Application

RPMI-1640 Medium has been used to culture murine fibroblast cells.[1] It has also been used to culture human ovarian and rat substantia nigra cells.[2]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Granados et al.
Journal of cardiovascular translational research, 10(4), 374-390 (2017-05-04)
Decellularized scaffolds represent a promising alternative for mitral valve (MV) replacement. This work developed and characterized a protocol for the decellularization of whole MVs. Porcine MVs were decellularized with 0.5% (w/v) SDS and 0.5% (w/v) SD and sterilized with 0.1%
Andres Binolfi et al.
Nature communications, 7, 10251-10251 (2016-01-26)
Cellular oxidative stress serves as a common denominator in many neurodegenerative disorders, including Parkinson's disease. Here we use in-cell NMR spectroscopy to study the fate of the oxidation-damaged Parkinson's disease protein alpha-synuclein (α-Syn) in non-neuronal and neuronal mammalian cells. Specifically
Julia Huetter et al.
Cancer immunology research, 8(7), 895-911 (2020-04-22)
The immunoglobulin-like domain containing receptor 2 (ILDR2), a type I transmembrane protein belonging to the B7 family of immunomodulatory receptors, has been described to induce an immunosuppressive effect on T-cell responses. Besides its expression in several nonlymphoid tissue types, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service