Skip to Content
Merck
All Photos(2)

Key Documents

F9682

Sigma-Aldrich

FSL-LEa(tri)

Synonym(s):

FSL-LEA(GALb3[Fa4]GLCNb)-SA1-L1

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$500.00

$500.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$500.00

About This Item

Empirical Formula (Hill Notation):
C70H125N3NaO25P
Molecular Weight:
1462.71
UNSPSC Code:
12352201
NACRES:
NA.25

$500.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98% (TLC)

Quality Level

form

solid

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

Application

All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. Cells modified with KODE constructs are known as kodecytes and usually maintain their normal vitality and functionality.

FSL-Lea(tri) has been specifically designed to create live cells (kodecytes) expressing blood group Lea antigens but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces. This product can be used to attach reproducible and controlled levels of blood group Lea antigens to erythrocytes. These kodecytes will react with most anti-Lea blood grouping reagents including polyclonal reagents. Lea-kodecytes will also react with monoclonal anti-Leb as these reagents also cross-react with Lea.
All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. FSL-Lea(tri) has been specifically designed to create live cells (kodecytes) expressing blood group Lea antigens but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces.

Other Notes

FSL-Lea(tri) is a KODE technology construct designed to label hydrophobic surfaces including living cells with Lewis blood group Lea trisaccharide. All KODE FSL constructs consist of three essential designable features: a functional component (F), a spacer (S), and a diacyl lipid (L). FSL-Lea(tri) is comprised of the Lewis blood group Lea trisaccharide Galβ(Fucα4)GlcNAcβ representing F, conjugated via an O(CH2)3NH spacer (SA1) to an activated adipate derivative of dioleoyl­phos­pha­tidyl­ethanolamine (L1).

Legal Information

KODE Constructs are supplied by KODE Biotech Materials Limited under license from KODE Biotech Limited.
KODE is a trademark of KODE Biotech Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service