F8506
N-Formyl-Met-Phe
≥97% (HPLC)
Synonym(s):
Neutrophil Dysfunction Test Peptide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H20N2O4S
CAS Number:
Molecular Weight:
324.40
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CSCCC(NC=O)C(=O)NC(Cc1ccccc1)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E Schöpf et al.
International archives of allergy and applied immunology, 82(3-4), 380-382 (1987-01-01)
Atopic dermatitis (AD) is characterized by many signs of immunodeficiency. Our interest was to investigate if there are also alterations of the chemiluminescence (CL) response of polymorphonuclear leukocytes (PMN) as a measure of the release of toxic oxygen radicals. Isolated
I R Cooke et al.
Cell and tissue research, 253(1), 69-76 (1988-07-01)
The distribution of FMRFamide-like immunoreactive neurons in the nervous system of the slug Limax maximus was studied using immunohistochemical methods. Approximately one thousand FMRFamide-like immunoreactive cell bodies were found in the central nervous system. Ranging between 15 micron and 200
L T Williams et al.
Proceedings of the National Academy of Sciences of the United States of America, 74(3), 1204-1208 (1977-03-01)
Synthetic N-formylmethionyl peptides are chemotactic attractants for human polymorphonuclear leukocytes. The well-defined structure-activity relationship of these peptides in eliciting a chemotactic response suggests that the interaction of the peptides with a specific cellular binding site may initiate chemotaxis. By using
D F Jadwin et al.
American journal of clinical pathology, 76(4), 395-402 (1981-10-01)
The described procedure permits assessment of the neutrophil shape change response to formyl-methionyl peptides added directly to EDTA anticoagulated whole blood. Response is monitored by counting the resultant number of "bipolar" neutrophil forms present on stained blood films. The preparation
Acetorphan and diarrhoea.
A T Cole et al.
Gut, 34(3), 432-432 (1993-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service