Skip to Content
Merck
All Photos(1)

Key Documents

F7751

Sigma-Aldrich

Fluoxymesterone

solid (photosensitive)

Synonym(s):

11β,17β-Dihydroxy-9α-fluoro-17α-methyl-4-androsten-3-one, 9α-Fluoro-11β,17β-dihydroxy-17α-methylandrost-4-en-3-one, 9α-Fluoro-11β-hydroxy-17α-methyltestosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29FO3
CAS Number:
Molecular Weight:
336.44
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

Porcine pancreas
bovine milk (other parts like skeletal muscle/bile/bone/connective tissue/skin)
microbial (Mycobacterium sp.)

form

solid (photosensitive)

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

solubility

H2O: ≤0.5 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 6.4 mg/mL
ethanol: 7.3 mg/mL

application(s)

forensics and toxicology
veterinary

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@](C)(O)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)CC[C@]34C

InChI

1S/C20H29FO3/c1-17-8-6-13(22)10-12(17)4-5-15-14-7-9-19(3,24)18(14,2)11-16(23)20(15,17)21/h10,14-16,23-24H,4-9,11H2,1-3H3/t14-,15-,16-,17-,18-,19-,20-/m0/s1

InChI key

YLRFCQOZQXIBAB-RBZZARIASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fluoxymesterone is an androgen and anabolic steroid.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Fürstenberger et al.
Toxicological sciences : an official journal of the Society of Toxicology, 126(2), 353-361 (2012-01-26)
Anabolic androgenic steroids (AAS) are testosterone derivatives used either clinically, in elite sports, or for body shaping with the goal to increase muscle size and strength. Clinically developed compounds and nonclinically tested designer steroids often marketed as food supplements are
J E Olson et al.
Cancer, 79(6), 1138-1149 (1997-03-15)
The purpose of this study was to test the role of radiotherapy following total mastectomy, axillary dissection, and adjuvant systemic therapy in the management of operable locally advanced breast carcinoma. After undergoing mastectomy and axillary dissection, 426 patients with locally
Jeffrey N Miner et al.
Endocrinology, 148(1), 363-373 (2006-10-07)
A number of conditions, including osteoporosis, frailty, and sexual dysfunction in both men and women have been improved using androgens. However, androgens are not widely used for these indications because of the side effects associated with these drugs. We describe
Toktam Zohoorian-Abootorabi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 177-191 (2012-01-06)
This study was designed to examine the interaction of two anti-breast cancer drugs, i.e., fluoxymesterone (FLU) and cyclophosphamide (CYC), with human serum albumin (HSA) using different kinds of spectroscopic, zeta potential and molecular modeling techniques under imitated physiological conditions. The
G Falkson et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 16(5), 1669-1676 (1998-05-20)
To investigate the value of maintenance treatment for patients with metastatic breast cancer whose disease is in complete remission (CR). One hundred ninety-five women (141 eligible) whose disease was in CR or in CR except for bone metastases following six

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service