Skip to Content
Merck

Skip To

F7680

FAK (393-698), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FADK, FAK1, PTK2, pp125FAK

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352204
Specific activity:
221-299 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

221-299 nmol/min·mg

mol wt

~35 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTK2(5747)

General description

Focal adhesion kinase (FAK) or PTK2 (protein tyrosine kinase 2) is a tyrosine kinase, which resides in the cytoplasm, and concentrated at cellular focal points. It is a highly conserved 125kDa protein, which has a wide range of tissue expression. It is composed of 1,052 amino acids, and contains N- and C-termini flanking the central kinase domain. The N-terminal called the FERM (four-point-one, ezrin, radixin, moesin) domain, controls FAK activity, whereas the C-terminal contains the focal-adhesion-targeting (FAT) domain.[1]

Biochem/physiol Actions

Focal adhesion kinase (FAK) or PTK2 protein tyrosine kinase 2 plays multiple roles such as, control of transcription, cell cycle progression, apoptosis, cell migration, and metastasis in case of tumor cells. This attributes to its predominant role in cell signaling. It is involved in the infection of rabies virus (RABV).[1] Studies in Spanish population show that polymorphisms rs7460 and rs7843014 in this gene might be linked with exceptional longevity.[2] Phosphatidylinositol 4-phosphate 5-kinase type Iγ enzyme produces phosphatidylinositol-4,5-bisphosphate [PI(4,5)P2] in focal adhesion, which cross-links FAK activation to integrin signaling.[3]

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters