Skip to Content
Merck

Skip To

F6380

Anti-Human IgG (γ-chain specific)−FITC antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.5 mL
Check Cart for Availability
$133.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
5

$133.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:32

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Human IgG (γ-chain specific)-FITC antibody is specific for human IgG subclasses. Whole antiserum is fractionated and then further purified by ion exchange chromatography to provide the IgG fraction of antiserum. Goat anti-human IgG is conjugated to Sigma Fluorescein Isothiocyanate (FITC).

Immunogen

Pooled normal human serum

Application

Anti-Human IgG (γ-chain specific)-FITC antibodymay be used for immunofluorescent labelling of human peripheral blood lymphocytes at a working antibody dilution of 1:32 or 1:130. was used to determine IgG isotypes on human RBC by exhaustive photon reassignment microscopy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0132F9512F6257
technique(s)

direct immunofluorescence: 1:32

technique(s)

direct immunofluorescence: 1:32

technique(s)

immunofluorescence: 1:64-1:128 using Hep2 cells

technique(s)

direct immunofluorescence: 1:32

biological source

goat

biological source

goat

biological source

goat

biological source

sheep

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters