Skip to Content
Merck
All Photos(1)

Key Documents

F6178

Sigma-Aldrich

Fetal Bovine Serum

USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
biological source:
bovine fetus
origin:
USA origin
sterility:
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

sterility

sterile-filtered

composition

Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤25 mg/dL

origin

USA origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

≤25 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Application

FBS sourced in the United States is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bon-Kyoung Koo et al.
Nature, 488(7413), 665-669 (2012-08-17)
LGR5+ stem cells reside at crypt bottoms, intermingled with Paneth cells that provide Wnt, Notch and epidermal growth factor signals. Here we find that the related RNF43 and ZNRF3 transmembrane E3 ubiquitin ligases are uniquely expressed in LGR5+ stem cells.
null
Martina Belli et al.
Endocrinology, 159(4), 1690-1703 (2018-02-23)
Germline knockout studies in female mice demonstrated an essential role for forkhead box L2 (FOXL2) in early follicle development, whereas an inducible granulosa cell (GC)-specific deletion of Foxl2 in adults has shown ovary-to-testis somatic sex reprogramming. In women, over 120
Li Ding et al.
Autophagy, 13(7), 1176-1190 (2017-06-09)
Although the treatments of malignant glioma include surgery, radiotherapy and chemotherapy by oral drug administration, the prognosis of patients with glioma remains very poor. We developed a polyethylene glycol-dipalmitoylphosphatidyle- thanoiamine (mPEG-DPPE) calcium phosphate nanoparticles (NPs) injectable thermoresponsive hydrogel (nanocomposite gel)
Y Zhou et al.
Oncogene, 36(6), 731-745 (2016-08-02)
microRNA (miRNA) dysregulation is a common feature of cancer cells, but the complex roles of miRNAs in cancer are not fully elucidated. Here, we used functional genomics to identify oncogenic miRNAs in non-small cell lung cancer and evaluate their impact

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Protocols

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service