F5388
Carestream® BioMax® XAR Film
Synonym(s):
autoradiography film, xray film
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105341
Recommended Products
packaging
pkg of 50 ea bulk packaged
size
5 in. (13 cm) × 7 in. (18 cm)
Looking for similar products? Visit Product Comparison Guide
General description
- Coated with emulsion on both sides of the clear base.
- Recommended for chemiluminescence.
- Very versatile for autoradiography with high or low energy isotopes.
- Sensitivity can be increased with intensifying screens when using low-energy isotopes.
- When handling film in a darkroom, Kodak LED Safelight Darkroom Illuminator is recommended.
- All the XAR films are the same, just packaged differently.
The XAR-5 are single sheets of film that are bulk packaged with paper between each sheet.
Footnote
Kodak X-OMAT AR film was reformulated to become BioMax XAR film.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Eri Takeda et al.
PloS one, 6(11), e27815-e27815 (2011-11-24)
Vpr, an accessory protein of human immunodeficiency virus type 1, is a multifunctional protein that plays an important role in viral replication. We have previously shown that the region between residues 17 and 74 of Vpr (Vpr(N17C74)) contained a bona
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service