Skip to Content
Merck
All Photos(1)

Key Documents

F5132

Sigma-Aldrich

Factor II from human plasma

lyophilized powder, 5-20 units/mg protein

Synonym(s):

Prothrombin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

specific activity

5-20 units/mg protein

UniProt accession no.

foreign activity

factor X <0.01% of base activity
thrombin, none detected

storage temp.

−20°C

Gene Information

human ... F2(2147)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Plasma phospholipid binding protein; binds to endothelial cells, promotes clotting on the cell surfaces.

Unit Definition

One unit is the amount contained in one mL of normal human plasma.

Reconstitution

When reconstituted with 1 ml of deionized water, solution will contain the indicated activity in 0.5 M NaCl, 0.05 M Tris-HCl, pH 8.0.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayako Tomono et al.
Nihon Shokakibyo Gakkai zasshi = The Japanese journal of gastro-enterology, 110(5), 852-860 (2013-05-08)
A 78-year-old man was admitted for workup for a liver tumor. Both serum AFP and PIVKA-II levels were high (2260ng/ml and 806mAU/ml, respectively). Contrast-enhanced CT scan and MRI using Gd-EOB-DTPA demonstrated a liver tumor in segment 6 resembling the imaging
György Miklós Buzás
Orvosi hetilap, 154(21), 810-819 (2013-05-23)
Tumor markers are gene products which signal the occurrence of tumors in different organs as well as their response to surgery and chemotherapy. The discovery of tumor markers occurred after the demonstration of tumor-specific transplantation antigens in chemically or virally
Yuichiro Yanagida et al.
Surgery, 154(1), 48-57 (2013-05-21)
We tested the hypotheses that an increase in systemic thrombin activity occurs in both disseminated intravascular coagulation (DIC) with the fibrinolytic phenotype and in acute coagulopathy of trauma shock (ACoTS), and that the patients diagnosed as having ACoTS overlap or
Masahiko Tameda et al.
Cancer science, 104(6), 725-731 (2013-03-14)
Serum tumor markers, including α-fetoprotein (AFP) and des-γ-carboxy prothrombin (DCP), are currently used in the diagnosis of hepatocellular carcinoma (HCC). There is, however, an aberrant increase in serum DCP in patients with obstructive jaundice, vitamin K deficiency or who are
G V Papatheodoridis et al.
Journal of hepatology, 31(4), 747-750 (1999-11-07)
We evaluated the efficacy of recombinant factor VII to correct impaired haemostasis in a patient with liver cirrhosis requiring an invasive procedure. A test intravenous bolus of 80 microg/kg of recombinant factor VII was given to a Jehovah's Witness, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service