Skip to Content
Merck
All Photos(1)

Key Documents

F4928

Sigma-Aldrich

Flavin-Containing Monooxygenase-1, Microsomes human

recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution

Synonym(s):

FMO-1

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$580.00

$580.00


Estimated to ship onMay 14, 2025



Select a Size

Change View
1 VIAL
$580.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

$580.00


Estimated to ship onMay 14, 2025


recombinant

expressed in baculovirus infected insect cells

Quality Level

form

buffered aqueous solution

specific activity

See COA for Value

mol wt

55-60 kDa

packaging

vial of 2.5 mg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FMO1(2326)

Biochem/physiol Actions

The FMO′s are responsible for catalyzing the oxygenation of a variety of xenobiotic and drug compounds including environmental pollutants and carcinogens which contain a nucleophilic nitrogen, sulfur, selenium, or phosphorus-containing moiety associated with their respective oxides. These enzymes are classified as Phase I or oxidative drug metabolism enzymes.

Unit Definition

One unit will catalyze the production of 1.0 picomole of methyl p-tolylsulfoxide from methyl p-tolyl sulfide per minute at pH 9.5 at 37 deg C.

Physical form

Solution in 100 mM potassium phosphate buffer, pH 7.4.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service