Skip to Content
Merck

Skip To

F4259

2-Furaldehyde

Synonym(s):

Furfural

Sign In to View Organizational & Contract Pricing.

About This Item

EC Number:
202-627-7
UNSPSC Code:
12352200


density

1.16 g/mL at 25 °C (lit.)

storage temp.

2-8°C



signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

136.4 °F - closed cup

flash_point_c

58 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library