Skip to Content
Merck
All Photos(1)

Key Documents

F2177

Sigma-Aldrich

Follistatin 288 from mouse

>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym(s):

FS-288

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$910.00

$910.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$910.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$910.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

0.04-0.4 mg per mL

mol wt

calculated mol wt ~31 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Fst(14313)

General description

FST (follistatin) is an activin-binding protein, and exists in two isoforms due to alternate splicing, FS288 and FS315. It is a glycoprotein with a single chain, and acts as an activin antagonist. It has a high level of expression in fetal membranes and placenta.[1][2][3]

Biochem/physiol Actions

FST (follistatin) binds to and regulates activins, which in turn are TGF (transforming growth factor)-β superfamily members. The expression level of FST and its binding partner activin A is elevated in inflammatory disorders.[1] The activin A-follistatin system plays an essential role in the modulation of glucose and lipid metabolism, which might have an overall effect on fetal growth. In adipose tissue, it facilitates the adipogenic differentiation of progenitor cells.[3]
High affinity activin-binding protein that can act as an activin antagonist.

Physical form

Recombinant Mouse Follistatin 288 is supplied as a approximately 25 μg of protein lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA containing 1.25 mg of bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to neutralize activin-induced erythroid differentiation on K562 cells.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rita Linko et al.
BMC infectious diseases, 14, 253-253 (2014-06-03)
Activin A and its binding protein follistatin (FS) are increased in inflammatory disorders and sepsis. Overexpression of activin A in the lung causes similar histopathological changes as acute respiratory distress syndrome (ARDS). ARDS and severe respiratory failure are complications of
Silvia Näf et al.
PloS one, 9(4), e92175-e92175 (2014-04-26)
The Activin A-Follistatin system has emerged as an important regulator of lipid and glucose metabolism with possible repercussions on fetal growth. To analyze circulating activin A, follistatin and follistatin-like-3 (FSTL3) levels and their relationship with glucose metabolism in pregnant women
S Dasgupta et al.
Journal of postgraduate medicine, 58(3), 190-193 (2012-10-02)
Out of a panel of 37 candidate genes tested for linkage with polycystic ovary syndrome (PCOS), the strongest evidence of linkage was reported in the follistatin (FST) gene region. Subsequently, a couple of studies outside India investigated the FST gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service