Skip to Content
Merck
All Photos(1)

Key Documents

F1631

Sigma-Aldrich

Fast Violet B Salt

Dye content ≥97 %

Synonym(s):

4-Amino-5-methoxy-2-methylbenzanilide diazotated zinc double salt, 4-Benzoylamino-2-methoxy-5-methylbenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 41

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H14N3O2 · 0.5 ZnCl4
CAS Number:
Molecular Weight:
371.89
Colour Index Number:
37165
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ≥97%

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc(NC(=O)c2ccccc2)c(C)cc1[N+]#N.COc3cc(NC(=O)c4ccccc4)c(C)cc3[N+]#N

InChI

1S/2C15H13N3O2.4ClH.Zn/c2*1-10-8-13(18-16)14(20-2)9-12(10)17-15(19)11-6-4-3-5-7-11;;;;;/h2*3-9H,1-2H3;4*1H;/q;;;;;;+2/p-2

InChI key

KIWIFIXMWIMRBZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fast Violet B Salt is a diazonium salt. It is mainly required for cytochemical staining methods, for instance, alkaline phosphatase in leukocytes.

Application

Fast Violet B Salt has been used:
  • for alkaline phosphatase staining in cells
  • for hexosaminidase staining in cells
  • to study lysosomal membrane stability

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrated coastal monitoring of a gas processing plant using native and caged mussels.
Brooks S
The Science of the Total Environment, 426, 375-375 (2012)
Turgeon ML
Clinical Hematology: Theory and Procedures (2005)
Beta-glucuronidase and hexosaminidase are marker enzymes for different compartments of the endo-lysosomal system in mussel digestive cells.
Izagirre U
Cell and Tissue Research, 335, 441-441 (2009)
W Cockburn et al.
Analytical biochemistry, 189(1), 95-98 (1990-08-15)
A sensitive, quantitative assay for phosphenolpyruvate carboxylase which utilizes microtiter plates is described. The assay depends upon the production of a colored compound in the reaction between oxaloacetate, the product of the phosphoenolpyruvate reaction, and the dye Fast Violet B.
Jean Rivoal et al.
Analytical biochemistry, 300(1), 94-99 (2001-12-18)
We describe a method for the detection of isoforms of several glycolytic enzymes by activity staining after native PAGE. The staining is based on coupled enzyme assays carried out on the gel after electrophoresis and is linked to the disappearance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service