Skip to Content
Merck
All Photos(1)

Key Documents

F0509

Sigma-Aldrich

Fmoc-Phe-OH

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, Fmoc-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21NO4
CAS Number:
Molecular Weight:
387.43
Beilstein:
3597808
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

180-187 °C (lit.)

storage temp.

−20°C

SMILES string

OC(=O)[C@H](Cc1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H21NO4/c26-23(27)22(14-16-8-2-1-3-9-16)25-24(28)29-15-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h1-13,21-22H,14-15H2,(H,25,28)(H,26,27)/t22-/m0/s1

InChI key

SJVFAHZPLIXNDH-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service