All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C43H65N7O9
CAS Number:
Molecular Weight:
824.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CCCCC(NC=O)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(=O)NC(CCCC)C(=O)NC(Cc2ccc(O)cc2)C(=O)NC(CCCCN)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Potent chemoattractant for human neutrophils.
Radioiodinated molecule has full biological activity.
Radioiodinated molecule has full biological activity.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Djafarzadeh et al.
Experimental cell research, 236(2), 427-435 (1997-11-21)
We have studied activation-induced dephosphorylation of proteins in human neutrophils loaded with [32P]orthophosphate using two-dimensional gel electrophoresis and autoradiography. A major phosphoprotein of 20 kDa in resting neutrophils was markedly dephosphorylated upon activation of cells with chemotactic peptide or phorbol
G J Dobos et al.
The Clinical investigator, 72(5), 353-357 (1994-05-01)
We studied the effect of C5a pretreatment on phosphatidyl-inositol-4,5-bisphosphate (PIP2) hydrolysis and on the increase in peak and resting cytosolic calcium levels induced by C5a (0.1 and 10 nM) and/or N-formyl hexapeptide (FLPEP; 10 nM) in neutrophils isolated from patients
J F Hoffman et al.
Biochemistry, 35(40), 13047-13055 (1996-10-08)
With the aid of high time resolution kinetic data extracted from a flow cytometer, we determined that there are two N-formyl peptide receptor states for human neutrophils at 4 degrees C: a low affinity and a high affinity state. Competitive
S Seveau et al.
Journal of cell science, 110 ( Pt 13), 1465-1475 (1997-07-01)
We investigated a possible association of leukosialin (CD43), the major surface sialoglycoprotein of leukocytes, with neutrophil cytoskeleton. We first analysed the solubility of CD43 in Triton X-100 and observed that CD43 of resting neutrophils was mostly soluble. The small proportion
R McCoy et al.
The Journal of experimental medicine, 182(1), 207-217 (1995-07-01)
Although the classical chemotactic receptor for complement anaphylatoxin C5a has been associated with polymorphonuclear and mononuclear phagocytes, several recent studies have indicated that this receptor is expressed on nonmyeloid cells including human endothelial cells, vascular smooth muscle cells, bronchial and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service