Key Documents
EPS44I041005
Eppendorf® Mandatory stacking kit; Innova S44i, to allow installation
About This Item
UNSPSC Code:
41103800
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® S44I041005
Related Categories
General description
The Eppendorf Stacking Kit is designed for the double stacking of the Innova S44i shakers, enabling users to maximize lab space and increase productivity. This stacking kit provides a secure and stable platform for placing a second shaker on top of the first S44i unit. By utilizing this kit, researchers can optimize their work area without sacrificing performance. Easy to install and compatible with the S44i model, this stacking kit is an ideal solution for laboratories aiming to streamline their operations efficiently.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)