EPM1324-9906
Slide-Out Platform for Eppendorf® New Brunswick™ I26/I26R Incubator Shaker
for 250 mL Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum
About This Item
material
aluminum
feature
dedicated
for 250 mL Erlenmeyer flasks
non-Interchangeable
manufacturer/tradename
Eppendorf® M1324-9906 (NA)
size
76 cm × 46 cm
capacity
(40 x 250 mL Erlenmeyer flasks)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service