EPM1320-0004
Eppendorf® New Brunswick™ Innova® 43/43R Incubator Shaker
43R, refrigerated, diam. 2.5 cm (1 in.) , orbit, AC/DC input 120 V, 50 - 60 Hz (US)
About This Item
feature
20 °C below ambient to 80 °C temperature range
CO2 incubator with optional refrigeration
floor model
refrigerated
manufacturer/tradename
Eppendorf® M1320-0004 (NA)
parameter
(25-500 rpm with 2.5 cm orbit; 25-300 rpm with 5.1 cm orbit speed range)
±0.1 °C temp. stability (at 37 Celsius)
±0.25 °C (temperature uniformity at 37 Celcius)
W × D × H
115.6 cm (45.5 in.) × 81.3 cm (20 in.) × 103.1 cm (40.6 in.)
diam.
2.5 cm (1 in.) , orbit
platform size
76 cm × 46 cm
weight
152 kg
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Console style shaker/incubator with triple-eccentric drive – provides large capacity shaking for flasks up to 6 L
- Programmable Innova controls – automatically changes temperature, speed, and optional photosynthetic and UV germicidal lights at timed intervals
- Top mounted controls and display – for easy, ergonomic viewing and setting
- Foot pedal release with easy opening lid – provides hands-free operation
- RS-232 port for data logging
- Built-in water reservoir humidifies chamber to reduce sample evaporation while also protecting unit from spills and including a drain for easy cleaning
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service