Skip to Content
Merck
All Photos(1)

Key Documents

EPI018

Sigma-Aldrich

Sirtuin Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Activity Assay for Sirtuins, Fluorometric Sirtuin Kit, Sirtuin Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

shipped in

dry ice

storage temp.

−70°C

General description

Sirtuins are a class of proteins that possess either histone deacetylase or mono-ribosyltransferase activity. Sirtuins are localized in the cytoplasm, nucleus, nucleolus as well as mitochondria. They are associated with aging, cellular protection, sugar metabolism and cell cycle regulation. Unlike other known protein deacetylases, which simply hydrolyze acetyl-lysine residues, the sirtuin-mediated deacetylation reaction hydrolyzes acetyl-lysine and NAD. This hydrolysis yields the deacetylated substrate, O-acetyl-ADP-ribose and nicotinamide, itself an inhibitor of sirtuin activity. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. 

Suitability

Suitable for the detection of Sirtuin Activity in purified recombinant protein, cell and tissue lysate, nuclear extract, mitochondria and immunoprecipitated samples

Principle

In the Sirtuin Activity Assay Kit, the acetylated p53-AFC substrate is deacetylated by Sirtuins in the presence of NAD+ to generate the deacetylated p53-AFC substrate, nicotinamide and O-Acetyl-ADP Ribose. Cleavage of the deacetylated p53-AFC substrate by the Developer releases the fluorescent group, which is detected fluorometrically ((λex = 400/λem = 505 nm). HDAC′s also deacetylate the acetylated p53-AFC substrate. Trichostatin A is added to the reaction to specifically inhibit HDAC′s in samples. This kit provides a rapid, simple, sensitive, and reliable test to measure Sirtuin Activity in a variety of samples. The limit of quantification of the assay
is 0.06 μU of recombinant human SIRT6.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service