Skip to Content
Merck
All Photos(1)

Documents

EP951032085

Eppendorf® Deepwell plates, DNA LoBind, 96 wells

white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300

material

clear wells
colorless wells
conical bottom
polypropylene
white plate

sterility

non-sterile

feature

lid: no

packaging

pkg of 40 ea (5 bags × 8 plates)

manufacturer/tradename

Eppendorf® 951032085

capacity

500 μL

well volume

500 μL

color

blue border

suitability

suitable for PCR
suitable for molecular biology

Looking for similar products? Visit Product Comparison Guide

General description

Deepwell Plate 96/500µL, DNA LoBind, wells clear, 500 µL, LoBind, PCR clean, blue, 40 plates (5 bags × 8 plates)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly Hodge et al.
Journal of proteomics, 88, 92-103 (2013-03-19)
Mass spectrometry, in the past five years, has increased in speed, accuracy and use. With the ability of the mass spectrometers to identify increasing numbers of proteins the identification of undesirable peptides (those not from the protein sample) has also
Graziano Colombo et al.
Free radical biology & medicine, 52(9), 1584-1596 (2012-03-06)
Cigarette smoke, a complex mixture of over 7000 chemicals, contains many components capable of eliciting oxidative stress, which may induce smoking-related disorders, including oral cavity diseases. In this study, we investigated the effects of whole (mainstream) cigarette smoke on human
Cláudia P Grou et al.
The Journal of biological chemistry, 283(21), 14190-14197 (2008-03-25)
According to current models of peroxisomal biogenesis, newly synthesized peroxisomal matrix proteins are transported into the organelle by Pex5p. Pex5p recognizes these proteins in the cytosol, mediates their membrane translocation, and is exported back into the cytosol in an ATP-dependent
Jintang He et al.
Methods in molecular biology (Clifton, N.J.), 1002, 115-122 (2013-04-30)
Glycoproteins play an important role in cell signaling and cell-cell interaction. The alterations of glycoproteins are often relevant to progression of diseases, and these changed glycoproteins can be important biomarkers. The lectin-based glycoproteomic technology has extensively been used for high-throughput
Hongchang Qu et al.
Immunobiology, 218(4), 496-505 (2012-07-17)
Therapeutic modulation of the complement system has become increasingly important in line with the growing recognition of the role of complement in numerous diseases. Compstatin, a peptidic inhibitor that acts at the central level of the complement cascade, is currently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service