Skip to Content
Merck
All Photos(1)

Key Documents

EP951031801

Eppendorf® Deepwell Plate 96/500 uL

PCR Clean, volume 500 μL, white border with clear wells, pkg of 40 plates (5 bag x 8 plates)

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
volume:
500 μL
sterility:
non-sterile
feature:
PCR Clean
white border with clear wells
Pricing and availability is not currently available.

sterility

non-sterile

feature

PCR Clean
white border with clear wells

packaging

pkg of 40 plates (5 bag x 8 plates)

manufacturer/tradename

Eppendorf® 951031801 (North America)

technique(s)

PCR: suitable

volume

500 μL

General description

The Eppendorf® Deepwell Plate 96/500 uL is a high-performance plate designed for a wide range of manual and automated applications in the laboratory. It offers exceptional performance across various temperature conditions, from sample storage at -86 ºC to DNA denaturation at 100 ºC. The plate is constructed from the purest polypropylene (PP), ensuring high-quality and reliable results. These plates offer innovative features: more dependability, more efficiency, and more stability.

Application

Eppendorf® Deepwell Plate 96/500 uL is suitable for:

  • Sample storage and preparation
  • Bacteria and yeast cultivation
  • DNA and RNA isolation
  • Storage of cDNA or genomic banks
  • Storage of siRNA or oligonucleotide libraries
  • Storage and analysis of protein and DNA samples in Eppendorf LoBind® Plates
  • Oligonucleotide synthesis

Features and Benefits

  • OptiTrack® matrix: For enhanced well identification and reduced pipetting errors. The use of high-contrast alphanumeric labeling enables 30% faster well identification, leading to fewer mistakes during pipetting tasks.
  • Conical RecoverMax® well design: The good geometry is optimized to ensure maximum sample recovery and excellent mixing properties.
  • Minimized remaining/dead volume in automated applications: The matrix exhibits the highest uniformity from well to well, resulting in reduced dead volume.
  • Enhanced well sealing: The matrix features raised well rims and a smooth surface, ensuring reliable closing and compatibility with repeated heat-sealing methods.
  • g-safe® centrifugation stability: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • Barcode compatibility: The matrix is available with barcode options.
  • Purity options: The matrix is offered in two purity grades, namely, PCR clean (free of human DNA, DNase, RNase, and PCR inhibitors) and sterile (ensures it is free from pyrogens and sterile for sensitive applications).

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf LoBind is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service