Skip to Content
Merck
All Photos(1)

Key Documents

EP951020401

Eppendorf twin.tec® PCR Plates 96

skirted, size 150 μL, colorless, pkg of 25 ea

Synonym(s):

Twin.Tec 96-Well PCR Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
size:
150 μL
96 well
sterility:
non-sterile
feature:
skirt
skirted
Pricing and availability is not currently available.

sterility

non-sterile

feature

skirt
skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020401

technique(s)

PCR: suitable

size

150 μL
96 well

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Eppendorf twin.tec® PCR plates provide a reliable choice for every PCR assay. These plates offer a combination of purity, rigidity, and convenient features that simplify your daily laboratory routines. The robust polycarbonate frame ensures compatibility with automation and high-throughput processes. Additionally, the availability of different colors allows for easy organization of workflow steps. The raised well rims of twin.tec plates ensure efficient sealing, minimizing the potential for cross-contamination. Experience the benefits of twin.tec® plates and streamline your PCR experiments with ease and confidence.

Application

Eppendorf twin.tec® PCR Plates 96 is suitable for use in:

  • standard PCR
  • real-time PCR
  • high throughput PCR
  • small volume sample handling
PCR

Features and Benefits

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • The rim of the plate is designed to optimize barcode scanning and facilitate automated work processes.
  • The plate adheres to the recommended SBS footprint dimensions (127.76 × 85.47 mm ±0.25 mm).
  • Eight holes in the skirt help in the positioning of the plate and removal from the thermal cycler block.
  • The plate can be provided with a barcode upon request.
  • The plate is designed to be stackable, allowing for efficient use of space.
  • The low-profile design of the plate enhances PCR efficiency and enables optimal performance, especially with small sample volumes.
  • The plate has a maximum well volume of 150 µL when used with cap strips.
  • Suitable for quantitative real-time PCR as well-OptiTrack® matrix that enables faster sample identification and reduces pipetting errors.
  • Batch-tested and certified free of DNA, DNase, RNase, and PCR inhibitors (PCR clean).

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service