EP6731011515
Eppendorf® CellXpert® C170i CO2 Incubator
Copper interior, Oxygen control, inner door non-segmented, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Synonym(s):
Heaters
All Photos(1)
About This Item
UNSPSC Code:
41104405
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 6731011515
General description
CellXpert® C170i, non-segmented inner door, handle right side, 1 – 20 % oxygen control, copper package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Features and Benefits
- handle right side
- non-segmented inner door
- 180° HTD (standard with C170i)
- copper package: chamber, water tray and four shelves
- 100 – 127 V, 50 – 60 Hz
Legal Information
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)