EP6138000018
Eppendorf® μCuvette G1.0, microvolume measuring cell for Eppendorf BioPhotometer® and Eppendorf BioSpectrometer®
About This Item
Recommended Products
material
(quartz glass and aluminium)
feature
8.5 mm light path height
BSA concentration range: 75 ng/μL – 45.45 ng/μL
For colorimetric protein assays and florescence measurements
for use in Eppendorf BioPhotometer & Eppendorf BioSpectrometer
manufacturer/tradename
Eppendorf® 6138000018
volume measuring range
1.5-10 μL
W × D × H
12.5 mm × 12.5 mm × 48 mm
Features and Benefits
- Microvolume measuring cell for measuring 1.5 – 10 μL sample volumes
- Concentration determination of nucleic acids and proteins, amongst other target analytes
- Measurement of highly concentrated samples without prior dilution
- Suitable for fluorescence assays
- Low self-absorption (0.05 A at 260 nm)
- Hydrophobic surface coating on quartz glass for precise formation and positioning of the sample volume
- Simple cleaning to minimize the risk of sample carry-over
- A defined angle for folding the sides of the cuvette guarantees reproducible handling
- One-component hinged design prevents the loss of individual parts
- Exclusively available for use with Eppendorf BioPhotometer and Eppendorf BioSpectrometer
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service