EP5943000011
Eppendorf® Centrifuge 5910 Ri G
without Rotor, refrigerated, AC/DC input 230 V, 50—60 Hz, EuroPlug
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
refrigerated
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5943000011
Looking for similar products? Visit Product Comparison Guide
General description
Centrifuge 5910 Ri G, refrigerated, with touchscreen interface, without rotor, 230 V/50-60 Hz
Application
Application-Note_372_Centrifuge-5920-R_Centrifuge-5910-R_Faster-Isolation-of-GLP-centrifuge.pdf, , , , , ,
Features and Benefits
- Large 7-inch VisioNize® touch interface for super-fast parameter settings via unique favorites function
- User management with multiple authorization levels and documentation features supporting GLP/GxP requirements
- Swing-bucket rotors and adapters for tubes and bottles from 0.2 mL to 1,000 mL as well as all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 250 mL
- High centrifugation speed of up to 22,132 × g (14,000 rpm)
- Aerosol-tight Eppendorf QuickLock® caps and lids for fast and ergonomic lid locking
- Quiet operation to improve your work environment
- Centrifuge lid with soft-touch lid closure
- Temperature range from -11 °C to 40 °C
- Automatic rotor recognition and imbalance detection for maximum operational safety
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
VisioNize is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service