EP5804776003
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 1 tube 15 mL Falcon, pack of 2 ea
About This Item
Recommended Products
feature
holds 1 tube 15 mL Falcon
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804776003
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-34-6-38
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service