EP5702704008
Eppendorf® Fixed-angle rotor
F-35-30-17, 30 x 15 ml, incl. 20 adapters Falcon tubes and 30 rubber mats
About This Item
Recommended Products
feature
to hold 20 x 15 mL conical tubes
to hold 30 x 15 mL glass tubes
F-35-30-17, 30 x 15 ml, incl. 20 adapters Falcon tubes and 30 rubber mats
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5702704008
parameter
4,400 max. RCF (2,750 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/5702RH
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)