EP5322000008
Eppendorf® SmartBlock SmartExtender
Eppendorf® SmartBlocks for ThermoMixer C/Fx and ThermoStat C, vessel volume (1.5 mL)
Synonym(s):
Eppendorf® SmartExtender™
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5322000008
vessel volume
(1.5 mL)
compatibility
Eppendorf® SmartBlocks for ThermoMixer C/Fx and ThermoStat C
General description
Application
Features and Benefits
Up to 12 × 1.5 mL vessels
Temperature range from 3 °C above room temperature to 110 °C
Active heating only
Compatible with Eppendorf ThermoMixer C and Fx as well as ThermoStat C
Compatible to all Eppendorf SmartBlocks
Firmware update needed
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service