EP4924000088
Eppendorf® Reference® 2 G
single-channel, variable, volume 100-1,000 μL, blue, includes epT.I.P.S.® box
Synonym(s):
Adjustable volume pipette
About This Item
Recommended Products
feature
includes epT.I.P.S.® box
single-channel, variable
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4924000088
volume
100-1,000 μL
color
blue
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
- Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
- Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
- With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A step-by-step protocol for sample preparation, calibration, digestion, and inductively coupled plasma-mass spectrometric (ICP-MS) analysis of heavy metals in six different hemp-containing beverages.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service