EP4861000120
Eppendorf® Xplorer® 8-channel pipettor
volume 5-100 μL, multifunction rocker switch, yellow, for pipette tips 200 ul
About This Item
feature
for pipette tips 200 ul
multifunction rocker switch
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 4861000120
volume
5-100 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Multi-functional rocker with up is up and down is down functionality
- Selection dial for quick and easy selection of functions
- Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
- Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
- Powerful rechargeable battery for up to 8 hours work without the need for recharging
- Separate power socket: Work can continue while charging
- Adjustment to external conditions (e.g. geographic height, liquids with high density)
- Lower part of pipette can be autoclaved to ensure decontamination
- Spring-loaded tip cone (not for 1,200?μL multi-channel)
- Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service