EP022822993
Eppendorf® Vacufuge Plus Complete System
without rotor, incl. integrated pump, AC/DC input 120 V, 60 Hz, US 3-pin plug
Synonym(s):
Eppendorf Vacufuge, Vacufuge Plus System
About This Item
Recommended Products
feature
incl. integrated pump
without rotor
manufacturer/tradename
Eppendorf® 022822993 (US)
parameter
1400 rpm speed
technique(s)
cryopreservation: suitable
W × D × H
33 cm × 58 cm × 29 cm
weight
31.5 kg
Related Categories
General description
The Vacufuge plus is available with three configurations to meet the customers′ lab specific needs:
1) Basic Vacufuge for connecting to an existing vacuum pump
2) Complete system with an integrated solvent-resistant, maintenance and oil-free PTFE pump, and condensation trap
3) Complete system with vacuum port adds extra capability. It allows for using the integrated vacuum pump separately – without having to disconnect it from the system
- Choice of 3 operation modes (vacufuge, centrifuge, and desiccator) for a variety of application needs
- Choice of three application modes (aqueous, alcohol or high vapor pressure) to correspond with sample solvent—reduction of processing time up to 20%
- Choice of 4 heating levels (ambient, 30°C, 45°C, 60°C) allows safe and efficient concentration of various samples
- Brake function can optionally be turned off for use with delicate samples.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service