Skip to Content
Merck
All Photos(1)

Documents

EP022654306-1EA

Rotor for Eppendorf® 5430/5430R Centrifuges

F-35-6-30, non aerosol tight, lid, to hold, 6 x 15/50 mL conical tubes, to hold, 12 x Vacutainer® tubes

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

lid
to hold 12 x Vacutainer® tubes
to hold 6 x 15/50 mL conical tubes
F-35-6-30, non aerosol tight

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022654306 (North America)

parameter

7,830 max. RCF (7,745 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5430/5430R

Related Categories

General description

Eppendorf® centrifuges and microcentrifuges are utilized in various scientific disciplines such as microbiology, tissue culture, molecular biology, PCR, genomics, clinical applications, drug discovery, and proteomics. Eppendorf, a prominent company in the field of life sciences, specializes in the development and sale of a wide range of instruments, consumables, and services for liquid handling, sample handling, and cell handling in laboratories worldwide. Their product portfolio includes pipettes, automated pipetting systems, dispensers, centrifuges, mixers, and spectrometers. Eppendorf′s products find extensive use in both academic and commercial research laboratories, including those in the pharmaceutical, biotechnological, chemical, and food industries. We provide a diverse range of centrifuges, rotors, buckets, and adapters to cater to your specific processing requirements. This rotor is ideal for centrifuge models 5430/5430R. It comes with a rotor lid and six adapters each to hold 15 mL/50 mL conical tubes.

Application

Rotor for Eppendorf® 5430/5430R Centrifuges is suitable for centrifugation.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Vacutainer is a registered trademark of Becton-Dickinson & Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service