EP022637207
Rotor for Eppendorf ®5804/5804R and 5810/5810R Centrifuges
F-34-6-38, non aerosol tight, lid
Synonym(s):
Lid for Eppendorf® FA-45-48-11 Rotor
About This Item
Recommended Products
feature
lid
to hold 6 x 85 mL tubes
F-34-6-38, non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 002637207 (North America)
parameter
12000 ( 11,000) max. RCF (18,514 xg (15,557 xg )) (Model 5810 (model 5804/5804 R/5810))
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R and 5810/5810R
Related Categories
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service