Skip to Content
Merck
All Photos(1)

Key Documents

EP0030605900

Eppendorf® Microplate 96/V-PP, 96 wells

white polypropylene, conical bottom black wells, white border (wells black), PCR clean, polypropylene, pkg of 240 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom black wells
white polypropylene
size:
96 wells
Pricing and availability is not currently available.

material

conical bottom black wells
white polypropylene

packaging

pkg of 240 ea

manufacturer/tradename

Eppendorf® 0030605900

technique(s)

PCR: suitable

size

96 wells

Looking for similar products? Visit Product Comparison Guide

General description

Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
The “Eppendorf” Microplates are 96 well microplates made from high quality polypropylene material. It has been used extensively for various laboratory applications, such as sample storage, sample preparation, and various assays.

Application

The “Eppendorf” Microplates are suitable for a variety of applications including:
  • sample storage and preparation
  • assays requiring high resistance to temperature or solvents
  • storage of cDNA
  • active ingredient screening
  • combinatorial chemistry
  • protein analysis
  • genotyping
  • preparation of tissue and plant lysates
  • storage of oligonucleotide libraries

Features and Benefits

  • The [TM=“Eppendorf”] Microplates are made from high quality polypropylene, increasing the transparency and sample visibility.
  • The presence of raised well rims and a smooth surface supports effective sealing.
  • The [TM=“OptiTrack”] matrix feature enables faster sample identification and reduces pipetting error.
  • An optimized [TM="RecoverMax"] well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • Microplates have an exceptional centrifugation stability up to 6000 times gravity.
  • [TM=“Eppendorf”] Microplates can function effectively from -86 °C to 100 °C.
  • These microplates can be autoclaved at 121 °C for a duration of 20 minutes.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service