Key Documents
EP0030122283
Eppendorf® Protein LoBind tubes
new
capacity 25 mL, PCR clean, screw top cap, speed 17,000 x g rpm, pkg of 200 ea (4 bags x 50 tubes)
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
material
conical bottom
polypropylene
screw top cap
description
withstand 17,000xg
sterility
non-sterile
feature
DNase free
PCR clean
RNase free
graduations: no
lid
packaging
pkg of 200 ea (4 bags x 50 tubes)
manufacturer/tradename
Eppendorf® 0030122283
parameter
17,000 x g rpm speed
capacity
25 mL
color
colorless
suitability
suitable for PCR
binding type
Protein low binding
General description
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Protein LoBind Tubes, screw cap, Protein LoBind, 25 mL, PCR clean, colorless, 200 tubes (4 bags × 50 tubes)
Features and Benefits
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing to minimize evaporation
  • Rated up to 17,000_x_g centrifugation speed
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)