Skip to Content
Merck
All Photos(1)

Key Documents

EMU088481

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Stk3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTTGGAGAAGGGTCTTATGGAAGTGTTTTTAAAGCAATACATAAGGAATCTGGTCAAGTGGTTGCAATTAAGCAAGTACCTGTTGAGTCAGATCTTCAGGAAATAATCAAAGAAATTTCCATAATGCAACAATGTGACAGTCCATATGTTGTGAAGTACTATGGCAGTTACTTTAAGAACACAGACCTCTGGATTGTTATGGAGTACTGTGGAGCGGGGTCCGTTTCAGACATAATTAGATTGCGAAACAAGACATTAACAGAAGATGAAATTGCAACTATTCTAAAATCCACATTGAAAGGATTAGAATATTTGCATTTTATGAGGAAAATACACAGAGATATAAAAGCCGGGAATATTCTCCTCAATACAGAAGGACATGCAAAGCTTGCAGATTTTGGAGTGGCTGGCCAGTTAACAGATACAATGGCAAAACGCAACACTGTAATAGGAACCCCATTTTGGATGGCTCCTGAGGTAATTCAAGAAATAGGTTACAACTGTGTGGCTGACATCTGGTCCCTTGGCATTACTTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guiling Wang et al.
Oncotarget, 6(12), 9877-9886 (2015-04-19)
To date, microrchidia (MORC) family CW-type zinc-finger 2 (MORC2), has been found to be involved in p21-activated kinase1 (PAK1) pathway to maintain genomic integrity. Here, we explore its novel role in cancer. We demonstrate that PAK1-mediated MORC2 phosphorylation promotes cell
Juan Tang et al.
Oncotarget, 6(33), 34831-34845 (2015-10-27)
Oscillations in intracellular Ca2+ concentrations ([Ca2+]i) mediate various cellular function. Although it is known that [Ca2+]i oscillations are susceptible to dysregulation in tumors, the tumor-specific regulators of [Ca2+]i oscillations are poorly characterized. We discovered that CD147 promotes hepatocellular carcinoma (HCC)
Anna E Dart et al.
The Journal of cell biology, 211(4), 863-879 (2015-11-26)
P21-activated kinase 4 (PAK4) is a Cdc42 effector protein thought to regulate cell adhesion disassembly in a kinase-dependent manner. We found that PAK4 expression is significantly higher in high-grade human breast cancer patient samples, whereas depletion of PAK4 modifies cell
Alexandre K Rouquette-Jazdanian et al.
PloS one, 10(6), e0131823-e0131823 (2015-06-30)
Linker for Activation of T cells (LAT) is an adapter protein that is essential for T cell function. Knock-in mice with a LAT mutation impairing calcium flux develop a fatal CD4+ lymphoproliferative disease. miR-155 is a microRNA that is correlated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service