Skip to Content
Merck
All Photos(1)

Key Documents

EMU086511

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Thpo

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACAGCTGTCCCAAGCAGTACTTCTCAACTCCTCACACTAAACAAGTTCCCAAACAGGACTTCTGGATTGTTGGAGACGAACTTCAGTGTCACAGCCAGAACTGCTGGCCCTGGACTTCTGAGCAGGCTTCAGGGATTCAGAGTCAAGATTACTCCTGGTCAGCTAAATCAAACCTCCAGGTCCCCAGTCCAAATCTCTGGATACCTGAACAGGACACACGGACCTGTGAATGGAACTCATGGGCTCTTTGCTGGAACCTCACTTCAGACCCTGGAAGCCTCAGACATCTCGCCCGGAGCTTTCAACAAAGGCTCCCTGGCATTCAACCTCCAGGGTGGACTTCCTCCTTCTCCAAGCCTTGCTCCTGATGGACACACACCCTTCCCTCCTTCACCTGCCTTGCCCACCACCCATGGATCTCCACCCCAGCTCCACCCCCTGTTTCCTGACCCTTCCACCACCATGCCTAACTCTACCGCCCCTCATCCAGTCACAATGTACCCTCATCCCAGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lina M E Pettersson et al.
PloS one, 9(6), e100730-e100730 (2014-06-27)
Peripheral nerve injury results in dramatic upregulation in pituitary adenylate cyclase activating polypeptide (PACAP) expression in adult rat dorsal root ganglia and spinal motor neurons mirroring that described for the neurotrophin brain derived neurotrophic factor (BDNF). Thus, we posited that
Sae Hyun Park et al.
International journal of oncology, 44(3), 637-646 (2014-01-01)
Fascin1 (FSCN1) involved in cell motility and filopodia assembly plays important roles in biological processes such as cancer invasion and metastasis of multiple epithelial tumors. High-grade serous ovarian carcinoma (HGSOC) is aggressive and metastatic by acquiring an invasive phenotype and
Peng Zhang et al.
PloS one, 9(5), e97647-e97647 (2014-05-17)
Plasma kisspeptin levels dramatically increased during the first trimester of human pregnancy, which is similar to pregnancy specific glycoprotein-human chorionic gonadotropin. However, its particular role in the implantation and decidualization has not been fully unraveled. Here, the study was conducted
Linn-Karina M Selvik et al.
PloS one, 9(11), e112485-e112485 (2014-11-11)
Salt-inducible kinase 1 (SIK1/Snf1lk) belongs to the AMP-activated protein kinase (AMPK) family of kinases, all of which play major roles in regulating metabolism and cell growth. Recent studies have shown that reduced levels of SIK1 are associated with poor outcome
Shihai Liu et al.
International journal of clinical and experimental pathology, 7(8), 4857-4866 (2014-09-10)
Glioblastoma tumor cells release microvesicles, which contain mRNA, miRNA and angiogenic proteins. These tumor-derived microvesicles transfer genetic information and proteins to normal cells. Previous reports demonstrated that the increased microvesicles in cerebrospinal fluid (CSF) of patients with glioblastoma up-regulate procoagulant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service