Skip to Content
Merck
All Photos(1)

Documents

EMU077571

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cd74

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGACCCAGGACCATGTGATGCATCTGCTCACGAGGTCTGGACCCCTGGAGTACCCGCAGCTGAAGGGGACCTTCCCAGAGAATCTGAAGCATCTTAAGAACTCCATGGATGGCGTGAACTGGAAGATCTTCGAGAGCTGGATGAAGCAGTGGCTCTTGTTTGAGATGAGCAAGAACTCCCTGGAGGAGAAGAAGCCCACAGAGGCTCCACCTAAAGAGCCACTGGACATGGAAGACCTATCTTCTGGCCTGGGAGTGACCAGGCAGGAACTGGGTCAAGTCACCCTGTGAAGACAGAGGCCAGCTCTGCACAGCAGCAGCGCCCCCTGCTCTCCTGTGCCTCAGCCCTTCTTATGTTCCCTGATGTCACACCCCACTTCCCGTCTCCCTGCACCCTGGGGCTTGAGACTGGTGTCTGTTTCATCGTCCCAGGACACGGCAAATGAAGTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin J Bowen et al.
Developmental biology, 407(1), 145-157 (2015-07-19)
Proper remodeling of the endocardial cushions into thin fibrous valves is essential for gestational progression and long-term function. This process involves dynamic interactions between resident cells and their local environment, much of which is not understood. In this study, we
Yeyou Liang et al.
Metabolism: clinical and experimental, 64(12), 1682-1693 (2015-10-13)
Evidence shows that both macrophage migration inhibitory factor (MIF) and GLUT4 glucose transporter are involved in diabetic cardiomyopathy (DCM), but it remains largely unknown whether and how MIF regulates GLUT4 expression in cardiomyocytes. The present study aims to investigate the
Tomas Baldassarre et al.
Molecular cancer research : MCR, 13(6), 1044-1055 (2015-03-19)
Triple-negative breast cancers (TNBCs) are highly aggressive cancers that lack targeted therapies. However, EGFR is frequently activated in a subset of TNBCs and represents a viable clinical target. Because the endocytic adaptor protein Endophilin A2 (SH3GL1/Endo II) has been implicated
Zheren Shao et al.
PloS one, 10(7), e0132655-e0132655 (2015-07-24)
Melanomas cause over 76% of skin cancer deaths annually. Phosphatidylinositol 3-kinase (PI3K)-AKT-mammalian target of rapamycin (mTOR) signaling pathway is important for melanoma initiation and progression. In the current study, we evaluated the potential anti-melanoma effect of VS-5584, a novel and
Alexandre K Rouquette-Jazdanian et al.
PloS one, 10(6), e0131823-e0131823 (2015-06-30)
Linker for Activation of T cells (LAT) is an adapter protein that is essential for T cell function. Knock-in mice with a LAT mutation impairing calcium flux develop a fatal CD4+ lymphoproliferative disease. miR-155 is a microRNA that is correlated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service