Skip to Content
Merck
All Photos(1)

Key Documents

EMU077021

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Atad2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAGCAAAATAGACTTGAAAGCAACATAGACTTGAAAAATAACTCAAGTTCCTCTAATATTGAGAATGAACTTGAAGAGCCTAAGGAAACTACGGAGGGCACAGAACTGAGAAAAGACAGGATTGTTTGCAGGGGTGATGCCTCTGCCTCCCAGGTCACAGACATTCCCGAAGACAGTGAATCAAAAGAAATGGATTTTCTGCGGATGACTCTAGCTAGAGGTTCCCAGGTAGAGCAGCAGGAGCTCATCAGTATGGAGCAGGCTCTGGCCATCCTCTCTCAGCCGACACCCTCACTTGTGCTGGACCACAAGCAGTTAACAAATATTTTGAAGACGGTTGTTAAAAAAAGTCAGAAATATAATATATTCCAACTGGAGAATTTGTATGCAGTAATCAGCCAGTGTATTTATGAGCACCGCAGGGACTATGACAAAACAGCGCTTGTTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le Zheng et al.
Oncology reports, 33(5), 2337-2344 (2015-03-31)
The ATPase family AAA domain-containing protein 2 (ATAD2) is associated with many cellular processes, such as cell proliferation, invasion and migration. However, the molecular biological function of the ATAD2 gene in cervical cancer is unclear. The purpose of this study
Wen-Jing Lu et al.
Oncotarget, 6(39), 41722-41735 (2015-10-27)
The ATPase family, AAA domain containing 2 (ATAD2) is highly expressed in multiple cancers. We aim to understand the clinical and biological significance of ATAD2 over-expression in hepatocellular carcinoma (HCC), as a means to validate it as a therapeutic target

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service