Skip to Content
Merck
All Photos(1)

Key Documents

EMU075181

Sigma-Aldrich

MISSION® esiRNA

targeting mouse E2f1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGGATCTGGAGACTGACCATCAGTACCTCGCTGGTAGCAGTGGGCCATTCCGGGGCAGAGGCCGCCACCCAGGGAAAGGTGTGAAATCTCCGGGGGAGAAGTCACGCTATGAAACCTCACTAAATCTGACCACCAAACGCTTCTTGGAGCTGCTGAGCCGCTCAGCTGACGGTGTCGTTGACCTGAACTGGGCAGCTGAGGTGCTGAAGGTGCAGAAACGGCGCATCTATGACATCACCAATGTCCTGGAGGGCATCCAGCTCATTGCCAAGAAGTCCAAGAATCATATCCAGTGGCTAGGCAGCCACACCATGGTGGGGATTGGTAAGCGGCTTGAAGGCCTGACCCAGGACCTGCAGCAACTGCAGGAGAGTGAGCAGCAGCTGGATCACCTGATGCACATCTGTACCACACAGCTGCAACTGCTTTCGGAGGACTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanghao Chen et al.
Journal of neuro-oncology, 120(1), 43-53 (2014-08-21)
MicroRNAs (miRNAs) have gained much attention due to their critical roles in diverse biological events, including tumorigenesis. In this study, we demonstrate that miR-136 is down-regulated in two cohorts of patients with glioma. Furthermore, the low-level expression of miR-136 is
Xiaolei Jiang et al.
PloS one, 10(6), e0127951-e0127951 (2015-06-04)
The E2F1 transcription factor regulates cell proliferation and apoptosis through the control of a considerable variety of target genes. Previous work has detailed the role of other transcription factors in mediating the specificity of E2F function. Here we identify the
T J Kaitu'u-Lino et al.
Placenta, 36(8), 932-937 (2015-07-07)
Preeclampsia is a serious complication of pregnancy for which there are no efficacious medical treatments. Soluble endoglin is as an anti-angiogenic factor that contributes to the pathogenesis of the disease, however little is known about its molecular regulation in placenta.
Ning-Ai Liu et al.
The Journal of clinical endocrinology and metabolism, 100(7), 2557-2564 (2015-05-06)
Cushing disease, due to pituitary corticotroph tumor ACTH hypersecretion, drives excess adrenal cortisol production with adverse morbidity and mortality. Loss of glucocorticoid negative feedback on the hypothalamic-pituitary-adrenal axis leads to autonomous transcription of the corticotroph precursor hormone proopiomelanocortin (POMC), consequent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service